Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Common gamma Chain/IL-2 R gamma Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB7843
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB7843 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB7843 Supplier R&D Systems Supplier No. MAB7843
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Common gamma Chain/IL-2 R gamma Monoclonal specifically detects Common gamma Chain/IL-2 R gamma in Mouse samples. It is validated for Western Blot.

Specifications

Antigen Common gamma Chain/IL-2 R gamma
Applications Western Blot
Classification Monoclonal
Clone 644337
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P34902
Gene Alias CD132, CD132 antigen, CIDX, combined immunodeficiency, X-linked, common cytokine receptor gamma chain, common gamma-chain, cytokine receptor common subunit gamma, gamma(c), IL-2 R gamma, IL-2 receptor subunit gamma, IL2R gamma, IL-2R subunit gamma, IL2RG, IL-2RG, IMD4, interleukin 2 receptor, gamma, Interleukin-2 receptor subunit gamma, p64, SCIDX, SCIDX1, severe combined immunodeficiency
Gene Symbols Il2rg
Host Species Rat
Immunogen S. frugiperda insect ovarian cell line Sf21-derived recombinant mouse Common gamma Chain/ IL-2 R gamma Ser25-Ala263 (predicted) Accession # P34902
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3561
Test Specificity Detects mouse Common gamma Chain/ IL-2 R gamma in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human Common gamma Chain, recombinant mouse (rm) IL-2 R alpha , or rmIL-2 R beta is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.