Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Common gamma Chain/IL-2 R gamma Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB7843SP
This item is not returnable.
View return policy
Description
Common gamma Chain/IL-2 R gamma Monoclonal specifically detects Common gamma Chain/IL-2 R gamma in Mouse samples. It is validated for Western Blot.Specifications
Common gamma Chain/IL-2 R gamma | |
Monoclonal | |
LYOPH | |
Western Blot 2 ug/mL | |
P34902 | |
Il2rg | |
S. frugiperda insect ovarian cell line Sf21-derived recombinant mouse Common gamma Chain/ IL-2 R gamma Ser25-Ala263 (predicted) Accession # P34902 | |
25 μg | |
Primary | |
Detects mouse Common gamma Chain/ IL-2 R gamma in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human Common gamma Chain, recombinant mouse (rm) IL-2 R alpha , or rmIL-2 R beta is observed. | |
Mouse | |
Purified |
Western Blot | |
644337 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD132, CD132 antigen, CIDX, combined immunodeficiency, X-linked, common cytokine receptor gamma chain, common gamma-chain, cytokine receptor common subunit gamma, gamma(c), IL-2 R gamma, IL-2 receptor subunit gamma, IL2R gamma, IL-2R subunit gamma, IL2RG, IL-2RG, IMD4, interleukin 2 receptor, gamma, Interleukin-2 receptor subunit gamma, p64, SCIDX, SCIDX1, severe combined immunodeficiency | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3561 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction