Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Mouse Complement C3a ELISA Kit (Colorimetric)
SDP

Catalog No. NBP270037
Click to view available options
Quantity:
1 Kit

ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.

The Mouse Complement C3a ELISA Kit (Colorimetric) measures mouse Complement C3a in serum, plasma and other biological fluids.

Specifications

Assay Range 0.16 - 10 ng/mL
Assay Sensitivity 0.10 ng/mL
Cross Reactivity This kit recognizes Mouse C3a in samples. No significant cross-reactivity or interference between Mouse C3a and analogues was observed.
Product Type ELISA Kit (Colorimetric)
Sample Type Biological Fluids, Plasma, Serum
Specificity Mouse Complement C3a ELISA Kit (Colorimetric) recognizes Mouse C3a in samples. No significant cross-reactivity or interference between Mouse C3a and analogues was observed.
For Use With (Application) ELISA
Gene ID (Entrez) 718
Gene Symbol C3
Host Species Mouse
Quantity 1 Kit
Research Discipline Adaptive Immunity, Apoptosis, Autoimmune Diseases, Diabetes Research, Immunology, Inflammation, Innate Immunity, Neuroscience, Signal Transduction, Stem Cells
Storage Requirements Storage of components varies. See protocol for specific instructions.
Target Complement C3a
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.