Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Complement Component C1rLP Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF3990
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF3990 100 μg
AF3990SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF3990 Supplier R&D Systems Supplier No. AF3990
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Complement Component C1rLP Polyclonal specifically detects Complement Component C1rLP in Mouse samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen Complement Component C1rLP
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL
Gene Accession No. AAP41417
Gene Alias C1RL, C1RL1, C1RL1complement C1r subcomponent-like protein, C1r-like protein, C1r-like serine protease analog protein, C1r-LPcomplement C1r-like proteinase, C1RLPEC 3.4.21.-, CLSPa, complement component 1, r subcomponent-like, EC 3.4.21, EC 3.4.21.41
Gene Symbols C1RL
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Complement Component C1rLP Ser18-Asp456 Accession # AAP41417
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51279
Test Specificity Detects mouse Complement Component C1rLP in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 40% cross-reactivity with recombinant human (rh) C1rLP is observed and less than 1% cross-reactivity with rhC1R and rhC1S is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.