Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Complement Component C5a Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB21501SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB21501SP 25 μg
MAB21501100 100 μg
MAB21501500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB21501SP Supplier R&D Systems Supplier No. MAB21501SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 2 publications

Complement Component C5a Monoclonal specifically detects Complement Component C5a in Mouse samples. It is validated for Western Blot, Neutralization, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen Complement Component C5a
Applications Western Blot, Neutralization, ELISA
Classification Monoclonal
Clone 295108
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Neutralization 0.1-0.5 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P06684
Gene Alias C5, C5A, HE
Gene Symbols Hc
Host Species Rat
Immunogen E. coli-derived recombinant mouse Complement Component C5a Asn679-Arg755 Accession # P06684
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 727
Test Specificity Detects mouse Complement Component C5a in Western blots. In Western blots, this antibody does not cross-react with recombinant mouse Complement Component C3d, recombinant human (rh) Complement Component C3a, or rhComplement Component C5a.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.