Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Complement Component C5a Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Supplier: R&D Systems BAF2150
This item is not returnable.
View return policy
Description
Complement Component C5a Polyclonal specifically detects Complement Component C5a in Mouse samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).Specifications
Complement Component C5a | |
Polyclonal | |
Biotin | |
P06684 | |
Hc | |
E. coli-derived recombinant mouse C5a (R&D Systems, Catalog # 2150-C5) Asn679-Arg755 Accession # P06684 | |
50 μg | |
Primary | |
Detects mouse Complement Component C5a in ELISAs and Western blots. In sandwich immunoassays, less than 0.05% cross-reactivity with rhC5a, rhC3a, rmC3d and rhAlpha-2 macroglobulin is observed. | |
Mouse | |
IgG |
Western Blot, ELISA | |
LYOPH | |
Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
C5, C5A, HE | |
Goat | |
Affinity Purified | |
RUO | |
727 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction