Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Complement Component C5aR1 Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB6467
This item is not returnable.
View return policy
Description
Complement Component C5aR1 Monoclonal specifically detects Complement Component C5aR1 in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
Complement Component C5aR1 | |
Monoclonal | |
LYOPH | |
Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
P30993 | |
C5AR1 | |
NS0 mouse myeloma cell line transfected with mouse Complement Component C5a R1 Accession # P30993 | |
100 μg | |
Primary | |
This antibody detects mouse Complement Component C5a R1 in direct ELISAs. | |
Mouse | |
Purified |
Flow Cytometry, CyTOF | |
583837 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
C5A, C5a anaphylatoxin chemotactic receptor, C5aR, C5a-R, C5AR1, C5ARC5a anaphylatoxin receptor, C5R1C5a ligand, CD88, CD88 antigen, complement component 5 receptor 1, complement component 5 receptor 1 (C5a ligand), complement component 5a receptor 1, Complement Component C5a R1 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
728 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction