Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Contactin-3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB6644SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB6644SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB6644SP Supplier R&D Systems Supplier No. MAB6644SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Contactin-3 Monoclonal specifically detects Contactin-3 in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.

Specifications

Antigen Contactin-3
Applications Western Blot, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 524518
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q07409
Gene Alias BIG-1, CNTN3, contactin 3 (plasmacytoma associated), Contactin3, PANG, PCS
Gene Symbols CNTN3
Host Species Rat
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse Contactin-3 Glu20-Thr1003 Accession # Q07409
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5067
Test Specificity Detects mouse Contactin-3 in direct ELISAS. In direct ELISAs, 100% cross-reactivity with recombinant human (rh) Contactin-3 and no cross-reactivity with recombinant mouse (rm) Contactin-1, -4, -6, rhContactin-2 or -5 is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.