Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse CRACC/SLAMF7 Alexa Fluor 594-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FA46281T100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FA46281T100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FA46281T100 Supplier R&D Systems Supplier No. FAB46281T100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

CRACC/SLAMF7 Monoclonal specifically detects CRACC/SLAMF7 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen CRACC/SLAMF7
Applications Flow Cytometry
Classification Monoclonal
Clone 520914
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 594
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. Q8BHK6
Gene Alias 19A, CD2 subset 1, CD2-like receptor activating cytotoxic cells, CD319, CD319 antigen, CRACCCD2-like receptor-activating cytotoxic cells, CS119A24 protein, Membrane protein FOAP-12, Novel Ly9, novel LY9 (lymphocyte antigen 9) like protein, Protein 19A, SLAM family member 7, SLAMF7
Gene Symbols SLAMF7
Host Species Rat
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse CRACC/SLAMF7 Ala22-Gly224 Accession # Q8BHK6
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57823
Test Specificity Detects mouse CRACC/SLAMF7 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human CRACC is observed.
Target Species Mouse
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2a
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.