Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CRACC/SLAMF7 Alexa Fluor 647-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems FAB46281R100UG
This item is not returnable.
View return policy
Description
CRACC/SLAMF7 Monoclonal specifically detects CRACC/SLAMF7 in Mouse samples. It is validated for Flow Cytometry.Specifications
CRACC/SLAMF7 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q8BHK6 | |
SLAMF7 | |
Chinese hamster ovary cell line CHO-derived recombinant mouse CRACC/SLAMF7 Ala22-Gly224 Accession # Q8BHK6 | |
RUO | |
57823 | |
Mouse | |
IgG2a |
Flow Cytometry | |
520914 | |
Alexa Fluor 647 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
19A, CD2 subset 1, CD2-like receptor activating cytotoxic cells, CD319, CD319 antigen, CRACCCD2-like receptor-activating cytotoxic cells, CS119A24 protein, Membrane protein FOAP-12, Novel Ly9, novel LY9 (lymphocyte antigen 9) like protein, Protein 19A, SLAM family member 7, SLAMF7 | |
Rat | |
100 μg | |
Primary | |
Detects mouse CRACC/SLAMF7 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human CRACC is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction