Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CREG Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF1697
This item is not returnable.
View return policy
Description
CREG Polyclonal specifically detects CREG in Mouse samples. It is validated for Western Blot.Specifications
CREG | |
Polyclonal | |
Unconjugated | |
NP_035934 | |
CREG1 | |
E. coli-derived recombinant mouse CREG Arg32-Gln220 Accession # NP_035934 | |
100 μg | |
Primary | |
Detects mouse CREG in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 5% cross-reactivity with recombinant human CREG is observed. | |
Mouse | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
cellular repressor of E1A-stimulated genes, cellular repressor of E1A-stimulated genes 1CREG, CREG1, protein CREG1 | |
Goat | |
Affinity Purified | |
RUO | |
8804 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction