Learn More
Mouse CREG Antibody, R&D Systems™
GREENER_CHOICE
Goat Polyclonal Antibody
Supplier:  R&D Systems AF1697SP
 View more versions of this product
Catalog No. AF1697SP
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
CREG Polyclonal specifically detects CREG in Mouse samples. It is validated for Western Blot.
Specifications
Specifications
CREG
Polyclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
cellular repressor of E1A-stimulated genes, cellular repressor of E1A-stimulated genes 1CREG, CREG1, protein CREG1
Goat
Affinity Purified
RUO
8804
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Western Blot
LYOPH
Western Blot 0.1 ug/mL
NP_035934
CREG1
E. coli-derived recombinant mouse CREG Arg32-Gln220 Accession # NP_035934
25 μg
Primary
Detects mouse CREG in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 5% cross-reactivity with recombinant human CREG is observed.
Mouse
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only