Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Cripto Alexa Fluor™ 750-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB1538S100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB1538S100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB1538S100 Supplier R&D Systems Supplier No. FAB1538S100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

CRIPTO Monoclonal specifically detects CRIPTO in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen CRIPTO
Applications Flow Cytometry
Classification Monoclonal
Clone 237603
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 750
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. P51865
Gene Alias CFC-2, CR, Cripto-1, Cripto-1 growth factor, CRIPTOCRGF, Epidermal growth factor-like cripto protein CR1, TDGF1, TDGF-1, teratocarcinoma-derived growth factor 1
Gene Symbols TDGF1
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Cripto Arg26-Gln150 Accession # P51865
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6997
Test Specificity Detects mouse Cripto in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) ART, recombinant mouse (rm) ART, rmArtemin, recombinant drosophila DPP, rhGDNF, recombinant rat (rr) GDNF, rhLAP, rhLatent TGF-beta 1, rhTGF-beta 2, rhLefty-A, rmLefty-1, rrMIS, rhNeurturin, rmNodal, rhTGF-alpha , rhTGF-beta 1, rhTGF-beta 1.2, rhTGF-beta 3, or recombinant amphibian TGF-beta 5 is observed.
Target Species Mouse
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2b
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.