Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Cripto Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB1538SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1538SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB1538SP Supplier R&D Systems Supplier No. MAB1538SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

CRIPTO Monoclonal specifically detects CRIPTO in Mouse samples. It is validated for Flow Cytometry, Blockade of Receptor-ligand Interaction, Immunocytochemistry, CyTOF-ready.

Specifications

Antigen CRIPTO
Applications Flow Cytometry, Blockade of Receptor-ligand Interaction, Immunocytochemistry, CyTOF
Classification Monoclonal
Clone 237603
Concentration LYOPH
Conjugate Unconjugated
Dilution Flow Cytometry 0.25 ug/10^6 cells, Blockade of Receptor-ligand Interaction, Immunocytochemistry 8-25 ug/mL, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P51865
Gene Alias CFC-2, CR, Cripto-1, Cripto-1 growth factor, CRIPTOCRGF, Epidermal growth factor-like cripto protein CR1, TDGF1, TDGF-1, teratocarcinoma-derived growth factor 1
Gene Symbols Tdgf1
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Cripto Arg26-Gln150 Accession # P51865
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6997
Test Specificity Detects mouse Cripto in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) ART, recombinant mouse (rm) ART, rmArtemin, recombinant drosophila DPP, rhGDNF, recombinant rat (rr) GDNF, rhLAP, rhLatent TGF-beta 1, rhTGF-beta 2, rhLefty-A, rmLefty-1, rrMIS, rhNeurturin, rmNodal, rhTGF-alpha , rhTGF-beta 1, rhTGF-beta 1.2, rhTGF-beta 3, or recombinant amphibian TGF-beta 5 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.