Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Cripto Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF1538
This item is not returnable.
View return policy
Description
CRIPTO Polyclonal specifically detects CRIPTO in Mouse samples. It is validated for Western Blot, Blockade of Receptor-ligand Interaction, Immunocytochemistry.Specifications
CRIPTO | |
Polyclonal | |
Unconjugated | |
P51865 | |
Tdgf1 | |
Mouse myeloma cell line NS0-derived recombinant mouse Cripto Arg26-Gln150 Accession # P51865 | |
100 μg | |
Primary | |
Detects mouse Cripto in direct ELISAs and Western blots. In direct ELISAs, approximately 15% cross-reactivity with recombinant human Cripto-1 is observed. | |
Mouse | |
IgG |
Western Blot, Blockade of Receptor-ligand Interaction, Immunocytochemistry | |
LYOPH | |
Western Blot 0.1 ug/mL, Blockade of Receptor-ligand Interaction, Immunocytochemistry 5-15 ug/mL | |
CFC-2, CR, Cripto-1, Cripto-1 growth factor, CRIPTOCRGF, Epidermal growth factor-like cripto protein CR1, TDGF1, TDGF-1, teratocarcinoma-derived growth factor 1 | |
Sheep | |
Affinity Purified | |
RUO | |
6997 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction