Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CTHRC1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF5960
This item is not returnable.
View return policy
Description
CTHRC1 Polyclonal specifically detects CTHRC1 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
CTHRC1 | |
Polyclonal | |
Unconjugated | |
Q3UAP6 | |
CTHRC1 | |
Mouse myeloma cell line NS0-derived recombinant mouse CTHRC1 Ser33-Lys245 Accession # Q3UAP6 | |
100 μg | |
Primary | |
Detects mouse CTHRC1 in direct ELISAs and Western blots. | |
Mouse | |
IgG |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
collagen triple helix repeat containing 1, collagen triple helix repeat-containing protein 1, NMTC1, Protein NMTC1 | |
Sheep | |
Affinity Purified | |
RUO | |
115908 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction