Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse CTHRC1 Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF5960
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF5960 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF5960 Supplier R&D Systems Supplier No. BAF5960
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

CTHRC1 Polyclonal specifically detects CTHRC1 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen CTHRC1
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot
Gene Accession No. Q3UAP6
Gene Alias collagen triple helix repeat containing 1, collagen triple helix repeat-containing protein 1, NMTC1, Protein NMTC1
Gene Symbols CTHRC1
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse CTHRC1 Ser33-Lys245 Accession # Q3UAP6
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 115908
Test Specificity Detects mouse CTHRC1 in Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.