Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CTLA-4 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Supplier: R&D Systems AF476SP
This item is not returnable.
View return policy
Description
CTLA-4 Polyclonal specifically detects CTLA-4 in Mouse samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).Specifications
CTLA-4 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD, CD152, CD152 antigen, CD152IDDM12, CD28, celiac disease 3, CELIAC3, CTLA4, CTLA-4, cytotoxic T-lymphocyte antigen 4, cytotoxic T-lymphocyte protein 4, Cytotoxic T-lymphocyte-associated antigen 4, cytotoxic T-lymphocyte-associated protein 4, cytotoxic T-lymphocyte-associated serine esterase-4, GRD4, GSE, ICOS, ligand and transmembrane spliced cytotoxic T lymphocyte associated antigen 4 | |
Goat | |
Affinity Purified | |
RUO | |
1493 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, ELISA | |
LYOPH | |
Western Blot 0.1 ug/mL, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL | |
Q6GTR6 | |
CTLA4 | |
Mouse myeloma cell line NS0-derived recombinant mouse CTLA-4 Ala37-Phe162 Accession # Q6GTR6 | |
25 μg | |
Primary | |
Detects mouse CTLA-4 in ELISAs and Western blots. In sandwich immunoassays, less than 1% cross-reactivity with recombinant human CTLA-4, recombinant mouse (rm) CD28, rmICOS, and rmPD-1 is observed. | |
Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction