Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse CX3CR1 Alexa Fluor 488-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB5825G100
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB5825G100 100 Tests
FAB5825G025 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. FAB5825G100 Supplier R&D Systems Supplier No. FAB5825G100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 4 publications

CX3CR1 Polyclonal specifically detects CX3CR1 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen CX3CR1
Applications Flow Cytometry
Classification Polyclonal
Conjugate Alexa Fluor 488
Dilution Flow Cytometry 5 uL/10^6 cells
Gene Accession No. Q9Z0D9
Gene Alias Beta chemokine receptor-like 1, CCRL1, chemokine (C-C) receptor-like 1, chemokine (C-X3-C motif) receptor 1, chemokine (C-X3-C) receptor 1, CMKbRL1, CMK-BRL1, CMK-BRL-1, CMKDR1, CX3C chemokine receptor 1, Fractalkine receptor, G protein-coupled receptor 13, GPR13G-protein coupled receptor 13, GPRV28, V28, V28C-X3-C CKR-1
Gene Symbols CX3CR1
Host Species Goat
Immunogen E. coli-derived recombinant mouse CX3CR1 Met1-Thr32, Leu92-Lys104, Thr169-Val196, Lys258-Leu274 Accession # Q9Z0D9
Purification Method Affinity Purified
Quantity 100 Tests
Primary or Secondary Primary
Gene ID (Entrez) 1524
Test Specificity Detects human and mouse CX3CR1 in direct ELISAs and Western blots.
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.