Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CX3CR1 PE-conjugated Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 8 publications
$221.50 - $502.50
Specifications
Antigen | CX3CR1 |
---|---|
Dilution | Flow Cytometry 10 uL/10^6 cells |
Applications | Flow Cytometry |
Classification | Polyclonal |
Conjugate | R-PE |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FAB5825P025
![]() |
R&D Systems
FAB5825P025 |
25 Tests |
Each for $221.50
|
|
|||||
FAB5825P100
![]() |
R&D Systems
FAB5825P100 |
100 Tests |
Each for $502.50
|
|
|||||
Description
CX3CR1 Polyclonal specifically detects CX3CR1 in Mouse samples. It is validated for Flow Cytometry.Specifications
CX3CR1 | |
Flow Cytometry | |
R-PE | |
Mouse | |
Beta chemokine receptor-like 1, CCRL1, chemokine (C-C) receptor-like 1, chemokine (C-X3-C motif) receptor 1, chemokine (C-X3-C) receptor 1, CMKbRL1, CMK-BRL1, CMK-BRL-1, CMKDR1, CX3C chemokine receptor 1, Fractalkine receptor, G protein-coupled receptor 13, GPR13G-protein coupled receptor 13, GPRV28, V28, V28C-X3-C CKR-1 | |
CX3CR1 | |
IgG | |
Affinity Purified | |
Detects mouse CX3CR1 in direct ELISAs. Stains mouse CX3CR1 transfectants but not irrelevant transfectants in flow cytometry. |
Flow Cytometry 10 uL/10^6 cells | |
Polyclonal | |
Goat | |
Q9Z0D9 | |
1524 | |
E. coli-derived recombinant mouse CX3CR1 Met1-Thr32, Leu92-Lys104, Thr169-Val196, Lys258-Leu274 Accession # Q9Z0D9 | |
Primary | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title