Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CXADR Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF2654
This item is not returnable.
View return policy
Description
CXADR Polyclonal specifically detects CXADR in Mouse samples. It is validated for Western Blot.Specifications
CXADR | |
Polyclonal | |
Biotin | |
P97792 | |
CXADR | |
Mouse myeloma cell line NS0-derived recombinant mouse CXADR Leu20-Gly237 Accession # P97792 | |
50 μg | |
Primary | |
Detects mouse CXADR in Western blots. In Western blots, approximately 35% cross-reactivity with recombinant human CXADR is observed. | |
Mouse | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
CAR10Coxsackievirus B-adenovirus receptor, CAR4/6, coxsackie virus and adenovirus receptor, coxsackie virus B receptor, coxsackievirus and adenovirus receptor, CVB3 binding protein, CVB3 BP, CVB3-binding protein, hCAR, HCVADR | |
Goat | |
Affinity Purified | |
RUO | |
1525 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction