Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CXCL1/GRO alpha/KC/CINC-1 Alexa Fluor 350-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Supplier: R&D Systems IC4532U100UG
This item is not returnable.
View return policy
Description
CXCL1/GRO alpha/KC/CINC-1 Monoclonal specifically detects CXCL1/GRO alpha/KC/CINC-1 in Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
CXCL1/GRO alpha/KC/CINC-1 | |
Monoclonal | |
Alexa Fluor 350 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
CINC1, CINC-1, FSP, GRO alpha, GRO1, GROa, KC, MGSA, MGSA-a, MGSA-alpha, NAP-3, SCYB1 | |
E. coli-derived recombinant mouse CXCL1/GRO alpha/KC/CINC-1 Asn29-Lys96 Accession # P12850 | |
RUO | |
2919 | |
Mouse | |
IgG |
Flow Cytometry | |
1174A | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
P12850 | |
Rabbit | |
100 μg | |
Primary | |
Detects mouse CXCL1/GRO alpha/KC/CINC-1 in direct ELISAs. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction