Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse CXCL1/GRO alpha/KC/CINC-1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB4532100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4532100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB4532100 Supplier R&D Systems Supplier No. MAB4532100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

CXCL1/GRO alpha/KC/CINC-1 Monoclonal specifically detects CXCL1/GRO alpha/KC/CINC-1 in Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen CXCL1/GRO alpha/KC/CINC-1
Applications Flow Cytometry
Classification Monoclonal
Clone 1174A
Conjugate Unconjugated
Dilution Intracellular Staining by Flow Cytometry 0.25-1 μg/106 cells
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide
Gene Accession No. P12850
Gene Alias CINC1, CINC-1, FSP, GRO alpha, GRO1, GROa, KC, MGSA, MGSA-a, MGSA-alpha, NAP-3, SCYB1
Gene Symbols Cxcl1
Host Species Rabbit
Immunogen E. coli-derived recombinant mouse CXCL1/GRO alpha/KC/CINC-1, Asn29-Lys96, Accession No. P12850
Purification Method Protein A or G purified from cell culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 2919.0
Test Specificity Detects mouse CXCL1/GRO alpha/KC/CINC-1 in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.