Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse CXCL1/GRO alpha/KC/CINC-1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB453500
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB453500 500 μg
MAB453SP 25 μg
MAB453100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB453500 Supplier R&D Systems Supplier No. MAB453500
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 46 publications

CXCL1/GRO alpha/KC/CINC-1 Monoclonal specifically detects CXCL1/GRO alpha/KC/CINC-1 in Mouse samples. It is validated for Western Blot, Neutralization, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen CXCL1/GRO alpha/KC/CINC-1
Applications Western Blot, Neutralization, ELISA
Classification Monoclonal
Clone 48415
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Neutralization 0.05-0.25 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P12850
Gene Alias CINC1, CINC-1, FSP, GRO alpha, GRO1, GROa, KC, MGSA, MGSA-a, MGSA-alpha, NAP-3, SCYB1
Gene Symbols Cxcl1
Host Species Rat
Immunogen E. coli-derived recombinant mouse CXCL1/GRO alpha/KC/CINC-1 Arg20-Lys96 Accession # P12850
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2919
Test Specificity Detects mouse CXCL1/GRO alpha/KC/CINC-1 in direct ELISAs and Western blots. Shows 100% cross-reactivity with recombinant human (rh) CXCL2, less than 40% cross-reactivity with recombinant rat (rr) CINC-1, and no cross-reactivity with rrCINC-3, rhCXCL3, rhCXCL5, rhCXCL6, or recombinant mouse CXCL6.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.