Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CXCL1/GRO alpha/KC/CINC-1 Antibody, R&D Systems™

Rat Monoclonal Antibody
$214.00 - $561.50
Specifications
| Antigen | CXCL1/GRO alpha/KC/CINC-1 |
|---|---|
| Clone | 48415R |
| Dilution | Western Blot 1 ug/mL, Neutralization 0.05-0.25 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | Western Blot, Neutralization, ELISA |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB453RSP
![]() |
R&D Systems
MAB453RSP |
25 μg |
Each for $214.00
|
|
|||||
MAB453R100
![]() |
R&D Systems
MAB453R100 |
100 μg |
Each for $561.50
|
|
|||||
Description
CXCL1/GRO alpha/KC/CINC-1 Monoclonal specifically detects CXCL1/GRO alpha/KC/CINC-1 in Mouse samples. It is validated for Western Blot, Neutralization, ELISA Capture (Matched Antibody Pair).Specifications
| CXCL1/GRO alpha/KC/CINC-1 | |
| Western Blot 1 ug/mL, Neutralization 0.05-0.25 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Supernatant | |
| Mouse | |
| P12850 | |
| 2919 | |
| IgG2a | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 48415R | |
| Western Blot, Neutralization, ELISA | |
| Unconjugated | |
| Rat | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| CINC1, CINC-1, FSP, GRO alpha, GRO1, GROa, KC, MGSA, MGSA-a, MGSA-alpha, NAP-3, SCYB1 | |
| E. coli-derived recombinant mouse CXCL-1/GRO alpha/KC/CINC-1 Arg20-Lys96 Accession # P12850 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse CXCL-1/GRO alpha/KC/CINC-1 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title