Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse CXCL10/IP-10/CRG-2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB466SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB466SP 25 μg
MAB466100 100 μg
MAB466500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB466SP Supplier R&D Systems Supplier No. MAB466SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 8 publications

CXCL10/IP-10/CRG-2 Monoclonal specifically detects CXCL10/IP-10/CRG-2 in Mouse samples. It is validated for Western Blot, Neutralization, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen CXCL10/IP-10/CRG-2
Applications Western Blot, Neutralization, ELISA
Classification Monoclonal
Clone 134013
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Neutralization 10-40 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P17515
Gene Alias C7, chemokine (C-X-C motif) ligand 10, crg-2, CRG2, gIP-10, IFI10, INP10, IP-10, mob-1, SCYB10
Gene Symbols Cxcl10
Host Species Rat
Immunogen E. coli-derived recombinant mouse CXCL10/IP-10/CRG-2 Ile22-Pro98 Accession # P17515
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3627
Test Specificity Detects mouse CXCL10/IP-10/CRG-2 in ELISAs and Western blots. In Western blots, this antibody does not cross-react with recombinant human (rh) CXCL1, 2, 3, 5, 6, 7, 9, 10, 11, 12/SDF-1 alpha , 12/SDF-1 beta , 13, rmCXCL1, 2, 6, 9, 11, 12/SDF-1 alpha , 13, rpCXCL8, rrCXCL1, 3/CINC-2 alpha , 3/CINC-2 beta
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.