Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CXCL11/I-TAC Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 2 publications
Supplier: R&D Systems MAB572
This item is not returnable.
View return policy
Description
CXCL11/I-TAC Monoclonal specifically detects CXCL11/I-TAC in Mouse samples. It is validated for Western Blot, Neutralization.Specifications
CXCL11/I-TAC | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
beta-R1, b-R1, chemokine (C-X-C motif) ligand 11, H174, H174IP9, Interferon gamma-inducible protein 9, Interferon-inducible T-cell alpha chemoattractant, IP-9member 11, ITAC, I-TAC, I-TACMGC102770, SCYB9B, small inducible cytokine subfamily B (Cys-X-Cys), member 9B, Small-inducible cytokine B11 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
6373 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Western Blot, Neutralization | |
131327 | |
Western Blot 1 ug/mL, Neutralization 0.4-2.0 ug/mL | |
Q9JHH5 | |
CXCL11 | |
E. coli-derived recombinant mouse CXCL11/I-TAC Phe22-Met100 Accession # Q9JHH5 | |
500 μg | |
Primary | |
Detects mouse CXCL11/I-TAC in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant mouse CXC1, 2, 6, 9, 10, CXCL12/SDF-1 alpha , 13, 14, recombinant human CXCL1, 2, 3, 5, 6, 7, 8, 9, 10, 11, CXCL12/SDF-1 alpha , CXCL12/SDF-1 beta , 13, 14, 15, recombinant rat CXCL1, 2, CXCL3/CINC2 alpha , CXCL3/CINC-2 beta , 5, or recombinant porcine CXCL8 is observed. | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction