Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CXCL11/I-TAC Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF572
This item is not returnable.
View return policy
Description
CXCL11/I-TAC Polyclonal specifically detects CXCL11/I-TAC in Mouse samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).Specifications
CXCL11/I-TAC | |
Polyclonal | |
Biotin | |
Q9JHH5 | |
CXCL11 | |
E. coli-derived recombinant mouse CXCL11/I-TAC Phe22-Met100 Accession # Q9JHH5 | |
50 μg | |
Primary | |
Detects mouse CXCL11/I-TAC in ELISAs and Western blots. In sandwich immunoassays, less than 0.2% cross-reactivity with recombinant human CCL11, recombinant mouse (rm) BLC, rmCRG-2, rmGCP-2, rmMIG, rmPF4, and rmSDF-1 alpha is observed. | |
Mouse | |
IgG |
Western Blot, ELISA | |
LYOPH | |
Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
beta-R1, b-R1, chemokine (C-X-C motif) ligand 11, H174, H174IP9, Interferon gamma-inducible protein 9, Interferon-inducible T-cell alpha chemoattractant, IP-9member 11, ITAC, I-TAC, I-TACMGC102770, SCYB9B, small inducible cytokine subfamily B (Cys-X-Cys), member 9B, Small-inducible cytokine B11 | |
Goat | |
Affinity Purified | |
RUO | |
6373 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction