Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse CXCL13/BLC/BCA-1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB4701500
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4701500 500 μg
MAB4701SP 25 μg
MAB4701100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB4701500 Supplier R&D Systems Supplier No. MAB4701500
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 2 publications

CXCL13/BLC/BCA-1 Monoclonal specifically detects CXCL13/BLC/BCA-1 in Mouse samples. It is validated for Western Blot, Neutralization.

Specifications

Antigen CXCL13/BLC/BCA-1
Applications Western Blot, Neutralization
Classification Monoclonal
Clone 143608
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Neutralization 12-30 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. O55038
Gene Alias ANGIE, ANGIE2, B cell-attracting chemokine 1, B lymphocyte chemoattractant, BCA1, BCA-1, BCA1B-cell chemoattractant, BCA-1CXC chemokine BLC, B-cell-homing chemokine (ligand for Burkitt's lymphoma receptor-1), BLC, BLCSmall-inducible cytokine B13, BLR1L, B-lymphocyte chemoattractant, chemokine (C-X-C motif) ligand 13, chemokine (C-X-C motif) ligand 13 (B-cell chemoattractant), C-X-C motif chemokine 13, SCYB13, SCYB13B-cell-attracting chemokine 1, small inducible cytokine B subfamily (Cys-X-Cys motif), member 13 (B-cellchemoattractant)
Gene Symbols CXCL13
Host Species Rat
Immunogen E. coli-derived recombinant mouse CXCL13/BLC/BCA-1 Ile22-Ala109 Accession # O55038
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10563
Test Specificity Detects mouse CXCL13/BLC/BCA-1 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant cotton rat CXCL10, recombinant human CXCL1, 2, 3, 4, 5, 6, 7, 8, 9, 10, 11, 12/SDF-1 alpha , 12/SDF-1 beta , 13, 16, recombinant mouse CXCL1, 2, 4, 6, 7, 9, 10, 11, 12/SDF-1 alpha , 15, 16, recombinant porcine CXCL8, recombinant rat CXCL1, 2, 3/CINC-2 alpha , rr3/CINC-2 beta , or 5 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.