Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CXCL14/BRAK Antibody, R&D Systems™

Sheep Polyclonal Antibody has been used in 2 publications
$200.50 - $549.50
Specifications
Antigen | CXCL14/BRAK |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
CXCL14/BRAK Polyclonal specifically detects CXCL14/BRAK in Mouse samples. It is validated for Western Blot.Specifications
CXCL14/BRAK | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Sheep | |
Mouse | |
BMAC, Bolekine, BRAK, BRAKKEC, chemokine (C-X-C motif) ligand 14, Chemokine BRAK, CXC-X3, member 14 (BRAK), MIP-2 gamma, MIP2G, MIP-2G, NJACKec, SCYB14MGC10687, Small-inducible cytokine B14 | |
CXCL14 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
Q9JHH7 | |
9547 | |
E. coli-derived recombinant mouse CXCL14/BRAK Ser23-Glu99 Accession # Q9JHH7 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse CXCL14/BRAK in direct ELISAs and Western blots. In direct ELISAs, approximately 15% cross-reactivity with recombinant human BRAK is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title