Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse CXCL15/Lungkine Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB442
Change view
Click to view available options
Quantity:
25 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB442 500 μg
MAB442SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB442 Supplier R&D Systems Supplier No. MAB442
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 2 publications

CXCL15/Lungkine Monoclonal specifically detects CXCL15/Lungkine in Mouse samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen CXCL15/Lungkine
Applications Western Blot, ELISA
Classification Monoclonal
Clone 96708
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9WVL7
Gene Alias lungkine, Scyb15, weche
Gene Symbols CXCL15
Host Species Rat
Immunogen E. coli-derived recombinant mouse CXCL15/Lungkine Gln26-Ala167 Accession # Q9WVL7
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20309
Test Specificity Detects mouse CXCL15/Lungkine in ELISAs and Western blots. In sandwich immunoassays, no cross-reactivity or interference with recombinant mouse (rm) CXCL13, recombinant human (rh) CXCL13, rhCXCL5, rhCXCL8, rmMIP-2, recombinant rat CINC-2 beta , rmCX3CL1, rmCCL12, or rhCCL15 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.