Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse CXCL15/Lungkine Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF442
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF442 100 μg
AF442SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF442 Supplier R&D Systems Supplier No. AF442
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

CXCL15/Lungkine Polyclonal specifically detects CXCL15/Lungkine in Mouse samples. It is validated for Western Blot.

Specifications

Antigen CXCL15/Lungkine
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL
Gene Accession No. Q9WVL7
Gene Alias lungkine, Scyb15, weche
Gene Symbols CXCL15
Host Species Goat
Immunogen E. coli-derived recombinant mouse CXCL15/Lungkine (R&D Systems, Catalog # 442-LK) Gln26-Ala167 Accession # Q9WVL7
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20309
Test Specificity Detects mouse CXCL15/Lungkine in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with rm6Ckine, rmC10, rrCINC-1, rmBLC, rmCRG-2, rmEotaxin, rmFractalkine, rmGCP-2, rmJE, rmKC, rmLymphotactin, rmMARC, rmMDC, rmMIG, rmMIP-1 alpha , rmMIP-1 beta , rmRANTES, rmSDF-1 alpha , rmTCA-3 and rmTeck is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.