Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CXCL2/GRO beta/MIP-2/CINC-3 Antibody, R&D Systems™

Rat Monoclonal Antibody
$214.00 - $2020.00
Specifications
Antigen | CXCL2/GRO beta/MIP-2/CINC-3 |
---|---|
Clone | 40605 |
Dilution | Western Blot 1 ug/mL, Neutralization 1-4 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
Applications | Western Blot, Neutralization, ELISA |
Classification | Monoclonal |
Description
CXCL2/GRO beta/MIP-2/CINC-3 Monoclonal specifically detects CXCL2/GRO beta/MIP-2/CINC-3 in Mouse samples. It is validated for Western Blot, Neutralization, ELISA Capture (Matched Antibody Pair).Specifications
CXCL2/GRO beta/MIP-2/CINC-3 | |
Western Blot 1 ug/mL, Neutralization 1-4 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
Monoclonal | |
Rat | |
Mouse | |
P10889 | |
2920 | |
E. coli-derived recombinant mouse CXCL2/GRO beta/MIP-2/CINC-3 Ala28-Asn100 Accession # P10889 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse CXCL2 in ELISAs and Western blots. In ELISAs, does not cross-react with recombinant human (rh) CXCL2/GRO beta , rhCXCL3/GRO gamma , or recombinant mouse CXCL1/KC. |
40605 | |
Western Blot, Neutralization, ELISA | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
chemokine (C-X-C motif) ligand 2, CINC-2a, CINC3, CINC-3, C-X-C Motif Chemokine 2, GRO beta, GRO2, GRO2 oncogene, GROb, Gro-beta, Growth-regulated protein beta, Macrophage inflammatory protein 2-alpha, melanoma growth stimulatory activity beta, MGSA beta, MGSA-b, MGSA-beta, MIP2, MIP-2, MIP-2a, MIP2A, MIP2-alpha, SCYB2 | |
CXCL2 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title