Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Mouse CXCL2/MIP-2 DuoSet ELISA
GSA_VASDP

Catalog No. DY45205 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
5 Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DY45205 5 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DY45205 Supplier R&D Systems™ Supplier No. DY45205
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Includes

Mouse MIP-2 Capture Antibody; Mouse MIP-2 Detection Antibody; Mouse MIP-2 Standard; Streptavidin-HRP

Mouse CXCL2/MIP-2 DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Mouse CXCL2/MIP-2 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies mouse CXCL2/GRO beta/MIP-2/CINC-3.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Quantity 5 Plates
Species Mouse
Conjugate Biotin
For Use With (Application) ELISA
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.