Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Mouse CXCL2 (MIP-2) Recombinant Protein

Click to view available options
Quantity:
10 μg
100 μg
25 μg
500 μg
Description
Produced by activated monocytes and neutrophils and expressed at sites of inflammation. Hematoregulatory chemokine, which, in vitro, suppresses hematopoietic progenitor cell proliferation. GRO-beta(5-73) shows a highly enhanced hematopoietic activity.

Specifications
Specifications
Accession Number | P10889 |
Concentration | 200 μg/mL |
For Use With (Application) | Control, ELISA, Western Blot |
Formulation | PBS with no preservative |
Gene ID (Entrez) | 20310 |
Molecular Weight (g/mol) | 7.8 kD |
Name | Mouse CXCL2 (aa 28-100) |
Quantity | 10 μg |
Source | E. coli |
Storage Requirements | -20°C |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction