Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CXCL3/GRO gamma/CINC-2/DCIP-1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF5568SP
This item is not returnable.
View return policy
Description
CXCL3/GRO gamma/CINC-2/DCIP-1 Polyclonal specifically detects CXCL3/GRO gamma/CINC-2/DCIP-1 in Mouse samples. It is validated for Western Blot, Neutralization.Specifications
CXCL3/GRO gamma/CINC-2/DCIP-1 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
chemokine (C-X-C motif) ligand 3, CINC-2, CINC-2b, C-X-C motif chemokine 3, Dcip1, Gm1960, GRO gamma, GRO3, GRO3 oncogene, GROg, GRO-gamma, GRO-gamma(1-73), Growth-regulated protein gamma, Macrophage inflammatory protein 2-beta, melanoma growth stimulatory activity gamma, MGSA gamma, MIP-2b, MIP2B, MIP2-beta, SCYB3 | |
Sheep | |
Affinity Purified | |
RUO | |
2921 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Neutralization | |
LYOPH | |
Western Blot 0.1 ug/mL, Neutralization 0.5-3 ug/mL | |
AAI17017 | |
Cxcl3 | |
E. coli-derived recombinant mouse CXCL3/GRO gamma/CINC-2/DCIP-1 Ala28-Ser100 Accession # AAI17017 | |
25 μg | |
Primary | |
Detects mouse CXCL3/GRO gamma/CINC-2/DCIP-1 in direct ELISAs and Western blots. In direct ELISAs, approximately 50% cross-reactivity with recombinant rat CINC-2 is observed. | |
Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction