Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CXCL4/PF4 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF595SP
This item is not returnable.
View return policy
Description
CXCL4/PF4 Polyclonal specifically detects CXCL4/PF4 in Mouse samples. It is validated for Western Blot, Simple Western.Specifications
CXCL4/PF4 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
chemokine (C-X-C motif) ligand 4, C-X-C motif chemokine 4, CXCL4iroplact, Iroplact, MGC138298, Oncostatin-A, PF4, PF-4, platelet factor 4, SCYB4oncostatin-A | |
Goat | |
Affinity Purified | |
RUO | |
5196 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL, Simple Western 25 ug/mL | |
Q9Z126 | |
PF4 | |
E. coli-derived recombinant mouse CXCL4/PF4 Val30-Ser105 Accession # Q9Z126 | |
25 μg | |
Primary | |
Detects mouse CXCL4/PF4 in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant human CXCL4 is observed. | |
Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction