Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse CXCL7/Thymus Chemokine-1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB1091
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1091 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB1091 Supplier R&D Systems Supplier No. MAB1091
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

CXCL7/Thymus Chemokine-1 Monoclonal specifically detects CXCL7/Thymus Chemokine-1 in Mouse samples. It is validated for Western Blot, Neutralization.

Specifications

Antigen CXCL7/Thymus Chemokine-1
Applications Western Blot, Neutralization
Classification Monoclonal
Clone 159742
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Neutralization 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9EQI5
Gene Symbols Ppbp
Host Species Rat
Immunogen E. coli-derived recombinant mouse CXCL7/Thymus Chemokine-1 Lys40-Tyr113 Accession # Q9EQI5
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5473
Test Specificity Detects mouse CXCL7/Thymus Chemokine-1 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) CXCL1, 2, 3, 4, 5, 6, 7, 8, 9, 10, 11, 12, 13, 16, recombinant mouse (rm) CXCL1, 4, 6, 9, 10, 11, 12, 13, 15, 16, recombinant rat (rr) CXCL1, 2, 3, 5, recombinant porcine CXCL8, or recombinant cotton rat CXCL10 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.