Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CXCL7/Thymus Chemokine-1 Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $655.00
Specifications
Antigen | CXCL7/Thymus Chemokine-1 |
---|---|
Clone | 159703 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
Applications | Western Blot, ELISA |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB10911SP
![]() |
R&D Systems
MAB10911SP |
25 μg |
Each for $200.50
|
|
|||||
MAB10911
![]() |
R&D Systems
MAB10911 |
500 μg |
Each for $655.00
|
|
|||||
Description
CXCL7/Thymus Chemokine-1 Monoclonal specifically detects CXCL7/Thymus Chemokine-1 in Mouse samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).Specifications
CXCL7/Thymus Chemokine-1 | |
LYOPH | |
Western Blot, ELISA | |
Unconjugated | |
Rat | |
Mouse | |
Q9EQI5 | |
Ppbp | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
159703 | |
Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
5473 | |
E. coli-derived recombinant mouse CXCL7/Thymus Chemokine-1 Lys40-Tyr113 Accession # Q9EQI5 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse CXCL7/Thymus Chemokine-1 in ELISAs and Western blots. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title