Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CXCL7/Thymus Chemokine-1 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF793
This item is not returnable.
View return policy
Description
CXCL7/Thymus Chemokine-1 Polyclonal specifically detects CXCL7/Thymus Chemokine-1 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, ELISA Detection (Matched Antibody Pair).Specifications
CXCL7/Thymus Chemokine-1 | |
Polyclonal | |
Biotin | |
AAG36786 | |
Goat | |
Affinity Purified | |
RUO | |
5473 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunohistochemistry, ELISA | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
Ppbp | |
E. coli-derived recombinant mouse CXCL7/Thymus Chemokine-1 Lys40-Tyr113 Accession # AAG36786 | |
50 μg | |
Primary | |
Detects mouse CXCL7/Thymus Chemokine-1 in ELISAs and Western blots. In sandwich ELISAs, less than 0.02% cross-reactivity with recombinant rat (rr) TCK-1, rrCINC-3, recombinant human GRO-beta , recombinant mouse (rm) BLC, rmCRG-2/IP-10, and rmMIG is observed. | |
Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction