Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CXCL9/MIG Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF492SP
This item is not returnable.
View return policy
Description
CXCL9/MIG Polyclonal specifically detects CXCL9/MIG in Mouse samples. It is validated for Neutralization, ELISA Capture (Matched Antibody Pair).Specifications
CXCL9/MIG | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
chemokine (C-X-C motif) ligand 9, CMK, crg-10, C-X-C motif chemokine 9, Gamma-interferon-induced monokine, Humig, MIG, MIGSmall-inducible cytokine B9, monokine induced by gamma interferon, SCYB9Monokine induced by interferon-gamma | |
Goat | |
Affinity Purified | |
RUO | |
4283 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Neutralization, ELISA | |
LYOPH | |
Western Blot 0.5 ug/mL, Neutralization 6-20 ug/mL, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL | |
P18340 | |
CXCL9 | |
E. coli-derived recombinant mouse CXCL9/MIG Accession # P18340 | |
25 μg | |
Primary | |
Detects mouse CXCL9/MIG in ELISAs and Western blots. In sandwich immunoassays, less than 0.5% cross-reactivity with recombinant human (rh) CXCL9/MIG is observed and less than 0.005% cross-reactivity with recombinant mouse MIP-2, recombinant rat CINC-1, rhGRO alpha , rhGRO beta , and rhGRO gamma is observed. | |
Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction