Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse CXCR2/IL-8RB Alexa Fluor 750-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB2164S100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB2164S100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB2164S100 Supplier R&D Systems Supplier No. FAB2164S100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

CXCR2/IL-8RB Monoclonal specifically detects CXCR2/IL-8RB in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen CXCR2/IL-8RB
Applications Flow Cytometry
Classification Monoclonal
Clone 242216
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 750
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. P35343
Gene Alias CD182, CD182 antigen, CDw128b, chemokine (C-X-C motif) receptor 2, chemokine (CXC) receptor 2, CMKAR2, C-X-C chemokine receptor type 2, CXC-R2, CXCR-2, CXCR2 gene for IL8 receptor type B, GRO/MGSA receptor, High affinity interleukin-8 receptor B, IL-8 RB, IL-8 receptor type 2, IL-8R B, IL8R2, IL8RA, IL8RB, interleukin 8 receptor B, interleukin 8 receptor type 2, interleukin-8 receptor type B
Gene Symbols CXCR2
Host Species Rat
Immunogen C6 rat glioma cell line transfected with mouse CXCR2 Met1-Leu359 Accession # P35343
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3579
Test Specificity Stains mouse CXCR2 transfected cells but not CXCR1 transfectants.
Target Species Mouse
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2a
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.