Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CXCR2/IL-8RB PE-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 22 publications
$174.60 - $444.00
Specifications
| Antigen | CXCR2/IL-8RB |
|---|---|
| Clone | 242216 |
| Dilution | Flow Cytometry 10 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FAB2164P025
![]() |
R&D Systems
FAB2164P025 |
25 Tests |
Each for $174.60
|
|
|||||
FAB2164P100
![]() |
R&D Systems
FAB2164P100 |
100 Tests |
Each for $444.00
|
|
|||||
Description
CXCR2/IL-8RB Monoclonal specifically detects CXCR2/IL-8RB in Mouse samples. It is validated for Flow Cytometry.Specifications
| CXCR2/IL-8RB | |
| Flow Cytometry 10 uL/10^6 cells | |
| Monoclonal | |
| Purified | |
| Mouse | |
| CD182, CD182 antigen, CDw128b, chemokine (C-X-C motif) receptor 2, chemokine (CXC) receptor 2, CMKAR2, C-X-C chemokine receptor type 2, CXC-R2, CXCR-2, CXCR2 gene for IL8 receptor type B, GRO/MGSA receptor, High affinity interleukin-8 receptor B, IL-8 RB, IL-8 receptor type 2, IL-8R B, IL8R2, IL8RA, IL8RB, interleukin 8 receptor B, interleukin 8 receptor type 2, interleukin-8 receptor type B | |
| CXCR2 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Stains mouse CXCR2-transfected cells but not CXCR1 transfectants. |
| 242216 | |
| Flow Cytometry | |
| R-PE | |
| Rat | |
| P35343 | |
| 3579 | |
| C6 rat glioma cell line transfected with mouse CXCR2 Met1-Leu359 Accession # P35343 | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title