Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CXCR3 Alexa Fluor™ 405-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems FAB1685V100UG
This item is not returnable.
View return policy
Description
CXCR3 Monoclonal specifically detects CXCR3 in Mouse samples. It is validated for Flow Cytometry.Specifications
CXCR3 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
AAC40163 | |
CXCR3 | |
CHO Chinese hamster ovary cell line transfected with mouse CXCR3 Met1-Leu367 Accession # AAC40163 | |
RUO | |
2833 | |
Mouse | |
IgG2a |
Flow Cytometry | |
220803 | |
Alexa Fluor 405 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
CD183, CD183 antigen, chemokine (C-X-C motif) receptor 3, chemokine (C-X-C) receptor 3, CKR-L2IP-10 receptor, CMKAR3, C-X-C chemokine receptor type 3, CXCR3, CXC-R3, CXCR-3, G protein-coupled receptor 9CD182, GPR9, GPR9Mig-R, Interferon-inducible protein 10 receptor, IP10 receptor, IP10-R, Mig receptor, MigR | |
Rat | |
100 μg | |
Primary | |
Detects mouse CXCR3. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction