Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse CXCR3 PE-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB1685P100
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB1685P100 100 Tests
FAB1685P025 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. FAB1685P100 Supplier R&D Systems Supplier No. FAB1685P100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

CXCR3 Monoclonal specifically detects CXCR3 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen CXCR3
Applications Flow Cytometry
Classification Monoclonal
Clone 220803
Conjugate R-PE
Dilution Flow Cytometry 10 uL/10^6 cells
Gene Accession No. AAC40163
Gene Alias CD183, CD183 antigen, chemokine (C-X-C motif) receptor 3, chemokine (C-X-C) receptor 3, CKR-L2IP-10 receptor, CMKAR3, C-X-C chemokine receptor type 3, CXCR3, CXC-R3, CXCR-3, G protein-coupled receptor 9CD182, GPR9, GPR9Mig-R, Interferon-inducible protein 10 receptor, IP10 receptor, IP10-R, Mig receptor, MigR
Gene Symbols CXCR3
Host Species Rat
Immunogen CHO Chinese hamster ovary cell line transfected with mouse CXCR3 Met1-Leu367 Accession # AAC40163
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Primary or Secondary Primary
Gene ID (Entrez) 2833
Test Specificity Detects mouse CXCR3.
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.