Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CXCR4 PerCP-conjugated Antibody, R&D Systems™
Rat Monoclonal Antibody
Specifications
| Antigen | CXCR4 |
|---|---|
| Clone | 247506 |
| Dilution | Flow Cytometry 10 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
FAB21651C02
|
R&D Systems
FAB21651C025 |
25 Tests | N/A | N/A | N/A | ||||
|
FAB21651C10
|
R&D Systems
FAB21651C100 |
100 Tests | N/A | N/A | N/A | ||||
Description
CXCR4 Monoclonal specifically detects CXCR4 in Mouse samples. It is validated for Flow Cytometry.Specifications
| CXCR4 | |
| Flow Cytometry 10 uL/10^6 cells | |
| Monoclonal | |
| Purified | |
| Mouse | |
| 7852 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant |
| 247506 | |
| Flow Cytometry | |
| PerCP | |
| Rat | |
| P70658 | |
| CXCR4 | |
| Primary | |
| Detects mouse CXCR4 transfectants. Does not stain irrelevant transfectants. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title