Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CXCR5 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF6198SP
This item is not returnable.
View return policy
Description
CXCR5 Polyclonal specifically detects CXCR5 in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
CXCR5 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
BLR1, BLR1MDR-15, Burkitt lymphoma receptor 1, Burkitt lymphoma receptor 1, GTP binding protein (chemokine (C-X-C motif)receptor 5), Burkitt lymphoma receptor 1, GTP-binding protein, CD185, CD185 antigen, chemokine (C-X-C motif) receptor 5, C-X-C chemokine receptor type 5, CXCR5, CXC-R5, MDR15CXCR-5, MGC117347, Monocyte-derived receptor 15 | |
Goat | |
Affinity Purified | |
RUO | |
643 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Flow Cytometry, CyTOF | |
LYOPH | |
Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
Q04683 | |
CXCR5 | |
E. coli-derived recombinant mouse CXCR5 Met1-Pro57, Glu112-Thr126, Lys191-Phe221, and Leu283-Ile306, joined by linker peptides Accession # Q04683 | |
25 μg | |
Primary | |
Detects mouse CXCR5 in direct ELISAs. In direct ELISAs, less than 1% cross-reactivity with recombinant mouse CXCR7 is observed. | |
Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction