Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CXCR7/RDC-1 Alexa Fluor 405-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems FAB7167V100UG
This item is not returnable.
View return policy
Description
CXCR7/RDC-1 Monoclonal specifically detects CXCR7/RDC-1 in Mouse samples. It is validated for Flow Cytometry.Specifications
| CXCR7/RDC-1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| P56485 | |
| CXCR7 | |
| E. coli-derived recombinant mouse CXCR7/RDC-1 extracellular N-terminus and loops Met1-Leu47, Ser103-Lys118, Lys184-Glu213, Leu274-Ala296 Accession # P56485 | |
| RUO | |
| 57007 | |
| Mouse | |
| IgM |
| Flow Cytometry | |
| 734110 | |
| Alexa Fluor 405 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| ACKR3, chemokine (C-X-C motif) receptor 7, Chemokine orphan receptor 1CMKOR1C-X-C chemokine receptor type 7, CMKOR1, CXCR7, CXC-R7, CXCR-7, G protein-coupled receptor, GPR159, GPR159G-protein coupled receptor 159, RDC-1, RDC1G-protein coupled receptor RDC1 homolog | |
| Rat | |
| 100 μg | |
| Primary | |
| Detects mouse CXCR7/RDC-1 in direct ELISAs. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction