Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CXCR7/RDC-1 Antibody, R&D Systems™

Sheep Polyclonal Antibody has been used in 2 publications
Supplier: R&D Systems AF4227
This item is not returnable.
View return policy
Description
CXCR7/RDC-1 Polyclonal specifically detects CXCR7/RDC-1 in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
CXCR7/RDC-1 | |
Polyclonal | |
Unconjugated | |
P56485 | |
CXCR7 | |
Mouse myeloma cell line NS0-derived recombinant mouse CXCR7 extracellular N-terminus and loops Met1-Leu47, Ser103-Lys118, Lys184-Glu213, and Leu274-Ala296 Accession # P56485 | |
100 μg | |
Primary | |
Detects CXCR7/RDC-1 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant mouse CXCR5 is observed. | |
Mouse | |
IgG |
Western Blot, Flow Cytometry, CyTOF | |
LYOPH | |
Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
ACKR3, chemokine (C-X-C motif) receptor 7, Chemokine orphan receptor 1CMKOR1C-X-C chemokine receptor type 7, CMKOR1, CXCR7, CXC-R7, CXCR-7, G protein-coupled receptor, GPR159, GPR159G-protein coupled receptor 159, RDC-1, RDC1G-protein coupled receptor RDC1 homolog | |
Sheep | |
Affinity Purified | |
RUO | |
57007 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction