Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Cyr61/CCN1 Antibody, R&D Systems™
GREENER_CHOICE

Numéro de catalogue. AF4055
Modifier l'affichage
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AF4055 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
missing translation for 'nonReturnableMessage' missing translation for 'returnPolicyLabel'
Numéro de catalogue. AF4055 Fournisseur R&D Systems Code fournisseur AF4055
Ajouter au panier
Ajouter au panier
missing translation for 'nonReturnableMessage' missing translation for 'returnPolicyLabel'

Sheep Polyclonal Antibody has been used in 4 publications

Cyr61/CCN1 Polyclonal specifically detects Cyr61/CCN1 in Mouse samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.

Spécifications

Antigen Cyr61/CCN1
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Simple Western 10 ug/mL, Immunohistochemistry 5-15 ug/mL
Gene Accession No. NP_034646
Gene Alias CCN family member 1, CCN1, CCN1IGF-binding protein 10, Cysteine-rich angiogenic inducer 61, cysteine-rich, angiogenic inducer, 61, GIG1, GIG1cysteine-rich heparin-binding protein 61, IBP-10, IGFBP-10, IGFBP10cysteine-rich, anigogenic inducer, 61, Insulin-like growth factor-binding protein 10, protein CYR61, Protein GIG1
Gene Symbols CYR61
Host Species Sheep
Immunogen E. coli-derived recombinant mouse Cyr61/CCN1 Asp176-Gly281 Accession # NP_034646
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3491
Test Specificity Detects mouse Cyr61/CCN1 in direct ELISAs and Western blots. In direct ELISAs, this antibody shows approximately 10% cross-reactivity with recombinant human Cyr61/CCN1.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Afficher plus de résultats Afficher moins de résultats
missing translation for 'productTitle'
missing translation for 'pleaseSelectIssue'

missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.